accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 170161-27-0

H3CH3CCH3OONHNNNOOCH3H3CH3COOCH3H3CCH3

CAS RN 170161-27-0

IUPAC Name: 1,4,8-tri-tert-butyl 1,4,8,11-tetraazacyclotetradecane-1,4,8-tricarboxylate
Synonyms: 1,4,8-tri-tert-butyl 1,4,8,11-tetraazacyclotetradecane-1,4,8-tricarboxylate
Molecular Weight (g/mol): 500.68
Molecular Formula: C25H48N4O6
InChi Key: FIPOUUYFPSMVMX-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCCN(CCN(CCCNCC1)C(=O)OC(C)(C)C)C(=O)OC(C)(C)C

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products