accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 2015-56-7

NH2POOO

CAS RN 2015-56-7

IUPAC Name: {[amino(phenoxy)phosphoryl]oxy}benzene
Synonyms: {[amino(phenoxy)phosphoryl]oxy}benzene
Molecular Weight (g/mol): 249.21
Molecular Formula: C12H12NO3P
InChi Key: QWMUDOFWQWBHFI-UHFFFAOYSA-N
SMILES: NP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products