CAS RN 2162982-11-6
CAS RN 2162982-11-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express BAY-218, MedChemExpress
MedChemExpress BAY-218 (AHR antagonist 1) is an aryl hydrocarbon receptor (AHR) antagonist extracted from patent WO2017202816A1, example 23, has an IC50 of 39.9 nM in human cell line.
