accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 220239-67-8

OHOOFFF

CAS RN 220239-67-8

IUPAC Name: 2-[2-(trifluoromethoxy)phenyl]acetic acid
Synonyms: [2-(trifluoromethoxy)phenyl]acetic acid
Molecular Weight (g/mol): 220.15
Molecular Formula: C9H7F3O3
InChi Key: QFLBBLZROQPTAI-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CC=C1OC(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products