CAS RN 2316820-94-5
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
1
av
1
Resultat
1
R&D Systems™ QM-FN-SO3
A Tocris product, part of R&D Systems. Brain penetrant, NIR aggregation-induced emission active probe for in vivo imaging of Aβ plaques