CAS RN 2719-15-5
CAS RN 2719-15-5
IUPAC Name:
N-(2-methyl-4-nitrophenyl)acetamide
Synonyms:
N-(2-methyl-4-nitrophenyl)acetamide
Molecular Weight (g/mol):
194.19
Molecular Formula:
C9H10N2O3
InChi Key:
JZEOVPGWIWSSAK-UHFFFAOYSA-N
SMILES:
CC(=O)NC1=CC=C(C=C1C)[N+]([O-])=O
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 2'-Methyl-4'-nitroacetanilide, 97%, Thermo Scientific™
CAS: 2719-15-5 Molecular Formula: C9H10N2O3 Molecular Weight (g/mol): 194.19 MDL Number: MFCD00033896 InChI Key: JZEOVPGWIWSSAK-UHFFFAOYSA-N Synonym: n-2-methyl-4-nitrophenyl acetamide,4'-nitro-o-acetotoluidide,n1-2-methyl-4-nitrophenyl acetamide,acetamide, n-2-methyl-4-nitrophenyl,2'-methyl-4'-nitroacetanilide,acetyl-p-nitro-o-toluidine,2-methyl-4-nitroacetanilide,o-acetotoluidide, 4'-nitro,acetamide,n-2-methyl-4-nitrophenyl,maybridge1_001434 PubChem CID: 75936 IUPAC Name: N-(2-methyl-4-nitrophenyl)acetamide SMILES: CC(=O)NC1=CC=C(C=C1C)[N+]([O-])=O