accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 36238-83-2

CH3OHNO

CAS RN 36238-83-2

IUPAC Name: 7-methyl-2,3-dihydro-1,2-benzoxazol-3-one
Synonyms: 7-methyl-2H-1,2-benzoxazol-3-one
Molecular Weight (g/mol): 149.15
Molecular Formula: C8H7NO2
InChi Key: JUNCQBPTXAQOSA-UHFFFAOYSA-N
SMILES: CC1=C2ONC(=O)C2=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products