accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 4045-24-3

H3CONH

CAS RN 4045-24-3

IUPAC Name: 4-methoxypiperidine
Synonyms: 4-methoxypiperidine
Molecular Weight (g/mol): 115.18
Molecular Formula: C6H13NO
InChi Key: ZEYSHALLPAKUHG-UHFFFAOYSA-N
SMILES: COC1CCNCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products