accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 40751-89-1

H3COOONOO

CAS RN 40751-89-1

IUPAC Name: 2-methoxy-5-nitrobenzoate
Synonyms: 2-methoxy-5-nitrobenzoate
Molecular Weight (g/mol): 196.14
Molecular Formula: C8H6NO5
InChi Key: CVZUPRDSNNBZLH-UHFFFAOYSA-M
SMILES: COC1=CC=C(C=C1C([O-])=O)[N+]([O-])=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products