CAS RN 40949-94-8
CAS RN 40949-94-8
IUPAC Name:
potassium bis(trimethylsilyl)azanide
Synonyms:
potassium bis(trimethylsilyl)amide
Molecular Weight (g/mol):
199.49
Molecular Formula:
C6H18KNSi2
InChi Key:
IUBQJLUDMLPAGT-UHFFFAOYSA-N
SMILES:
[K+].C[Si](C)(C)[N-][Si](C)(C)C
1
–
1
of
1
results
1
Thermo Scientific Acros Potassium bis(trimethylsilyl)amide, 0.7M solution in toluene, +99.4%, Thermo Scientific Chemicals
CAS: 40949-94-8 | C6H18KNSi2 | 199.485 g/mol