accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 42287-90-1

BrOHO

CAS RN 42287-90-1

IUPAC Name: 3-(3-bromophenyl)propanoic acid
Synonyms: 3-(3-bromophenyl)propanoic acid
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: DWKWMFSWLCIMKI-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC(Br)=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products