accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 4378-13-6

H3C(R)(S)OHNH2OOH3CCH3CH3

CAS RN 4378-13-6

IUPAC Name: 2-amino-3-(tert-butoxy)butanoic acid
Synonyms: 2-amino-3-(tert-butoxy)butanoic acid
Molecular Weight (g/mol): 175.23
Molecular Formula: C8H17NO3
InChi Key: NMJINEMBBQVPGY-UHFFFAOYNA-N
SMILES: CC(OC(C)(C)C)C(N)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products