CAS RN 454-67-1
CAS RN 454-67-1
IUPAC Name:
3-fluoro-5-(trifluoromethyl)aniline
Synonyms:
3-fluoro-5-(trifluoromethyl)aniline
Molecular Weight (g/mol):
179.12
Molecular Formula:
C7H5F4N
InChi Key:
WQKQODZOQAFYPR-UHFFFAOYSA-N
SMILES:
NC1=CC(F)=CC(=C1)C(F)(F)F
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 3-Fluoro-5-(trifluoromethyl)aniline, 97%, Thermo Scientific™
CAS: 454-67-1 Molecular Formula: C7H5F4N Molecular Weight (g/mol): 179.12 MDL Number: MFCD00061315 InChI Key: WQKQODZOQAFYPR-UHFFFAOYSA-N Synonym: 3-amino-5-fluorobenzotrifluoride,3-fluoro-5-trifluoromethyl aniline,3-fluoro-5-trifluoromethyl-phenylamine,3-fluoro-5-trifluorobenzenamine,3-fluoro-5-trifluoromethyl-aniline,5-fluoro-3-trifluoromethyl aniline,benzenamine, 3-fluoro-5-trifluoromethyl,3-fluoro-5-trifluoromethyl phenylamine,pubchem1541 PubChem CID: 2735913 IUPAC Name: 3-fluoro-5-(trifluoromethyl)aniline SMILES: NC1=CC(F)=CC(=C1)C(F)(F)F