accessibility menu, dialog, popup

UserName

Save up to 60% on cold storage essentials

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 5192-04-1

NH2HN

CAS RN 5192-04-1

IUPAC Name: 1H-indol-7-amine
Synonyms: 1H-indol-7-amine
Molecular Weight (g/mol): 132.17
Molecular Formula: C8H8N2
InChi Key: WTFWZOSMUGZKNZ-UHFFFAOYSA-N
SMILES: NC1=C2NC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products