accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 57-91-0

H3C(S)(S)(R)OH(S)(R)HO

CAS RN 57-91-0

IUPAC Name: (1R,3aS,3bR,9bS,11aS)-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1,7-diol
Synonyms: estradiol 17 α epiestradiol α-estradiol 1,3,5-estratriene-3,17a-diol 17a-oestradiol 17-epiestradiol oestradiol-17α oestradiol-17-α alfatradiol epiestrol
Molecular Weight (g/mol): 272.39
Molecular Formula: C18H24O2
InChi Key: VOXZDWNPVJITMN-SFFUCWETSA-N
SMILES: C[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1CC[C@H]2O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products