accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 6092-80-4

HClH2NO

CAS RN 6092-80-4

IUPAC Name: hydrogen O-phenylhydroxylamine chloride
Synonyms: hydrogen O-aminophenol chloride
Molecular Weight (g/mol): 145.59
Molecular Formula: C6H8ClNO
InChi Key: DBTXKJJSFWZJNS-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NOC1=CC=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products