CAS RN 78686-79-0
CAS RN 78686-79-0
IUPAC Name:
methyl 5-bromo-2-chloropyridine-3-carboxylate
Synonyms:
methyl 5-bromo-2-chloropyridine-3-carboxylate
Molecular Weight (g/mol):
250.48
Molecular Formula:
C7H5BrClNO2
InChi Key:
MOMQDEDQGJAKII-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(Br)=CN=C1Cl
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar Methyl 5-bromo-2-chloronicotinate, 98%
CAS: 78686-79-0 Molecular Formula: C7H5BrClNO2 Molecular Weight (g/mol): 250.476 MDL Number: MFCD03844848 InChI Key: MOMQDEDQGJAKII-UHFFFAOYSA-N PubChem CID: 2763343 IUPAC Name: methyl 5-bromo-2-chloropyridine-3-carboxylate SMILES: COC(=O)C1=CC(=CN=C1Cl)Br