CAS RN 80336-64-7
CAS RN 80336-64-7
IUPAC Name:
2-bromo-1-[4-(2-methylpropyl)phenyl]propan-1-one
Synonyms:
2-bromo-1-[4-(2-methylpropyl)phenyl]propan-1-one
Molecular Weight (g/mol):
269.18
Molecular Formula:
C13H17BrO
InChi Key:
SVGMFVCPJFHTRE-UHFFFAOYNA-N
SMILES:
CC(C)CC1=CC=C(C=C1)C(=O)C(C)Br
1
–
1
of
1
results
1
Thermo Scientific Maybridge 2-Bromo-1-(4-isobutylphenyl)propan-1-one, 97%, Thermo Scientific™
CAS: 80336-64-7 Molecular Formula: C13H17BrO Molecular Weight (g/mol): 269.18 MDL Number: MFCD00831700 InChI Key: SVGMFVCPJFHTRE-UHFFFAOYNA-N Synonym: 2-bromo-1-4-isobutylphenyl propan-1-one,2-bromo-1-4-2-methylpropyl phenyl propan-1-one,1-4-isobutylphenyl-2-bromo-1-propanone,2-bromo-4'-2-methylprop-1-yl propiophenone,2-bromo-1-4-2-methylpropyl phenyl-1-propanone,2-bromanyl-1-4-2-methylpropyl phenyl propan-1-one,1-propanone,2-bromo-1-4-1,1-dimethylethyl phenyl PubChem CID: 2735576 IUPAC Name: 2-bromo-1-[4-(2-methylpropyl)phenyl]propan-1-one SMILES: CC(C)CC1=CC=C(C=C1)C(=O)C(C)Br