accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 827-01-0

ClHNO

CAS RN 827-01-0

IUPAC Name: 5-chloro-1H-indole-3-carbaldehyde
Synonyms: 5-chloro-1H-indole-3-carbaldehyde
Molecular Weight (g/mol): 179.6
Molecular Formula: C9H6ClNO
InChi Key: YXEXOIGXNYITQH-UHFFFAOYSA-N
SMILES: ClC1=CC=C2NC=C(C=O)C2=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products