accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 850623-61-9

BOCH3FFFK

CAS RN 850623-61-9

IUPAC Name: potassium (4-butoxyphenyl)trifluoroboranuide
Synonyms: potassium (4-butoxyphenyl)trifluoroboranuide
Molecular Weight (g/mol): 256.12
Molecular Formula: C10H13BF3KO
InChi Key: UEVIEKRZQJRPGE-UHFFFAOYSA-N
SMILES: [K+].CCCCOC1=CC=C(C=C1)[B-](F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products