CAS RN 89464-63-1
IUPAC Namn:
methyl 2-(2-methoxy-2-oxoacetamido)acetate
Synonymer:
methyl 2-(2-methoxy-2-oxoacetamido)acetate
Molekylvikt (g/mol):
175.14
Molekylär sammansättning:
C6H9NO5
InChi Key:
BNJOZDZCRHCODO-UHFFFAOYSA-N
SMILES:
COC(=O)CNC(=O)C(=O)OC
1
–
2
av
2
Resultat
1
2
DMOG, MedChemExpress
MedChemExpress DMOG (Dimethyloxallyl Glycine) is a cell permeable and competitive inhibitor of HIF-PH, which results in HIF-1α stabilisation and accmulation in vitro and in vivo. DMOG is an α-ketoglutarate analogue and inhibits α-KG-dependent hydroxylases. DMOG acts as a pro-angiogenic agent and plays a protective role in experimental model of colitis and diarrhoea via HIF-1 related signal. DMOG induces cell autophagy.