accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 97-72-3

H3CCH3OOOCH3CH3

CAS RN 97-72-3

IUPAC Name: 2-methylpropanoyl 2-methylpropanoate
Synonyms: 2-methylpropanoyl 2-methylpropanoate
Molecular Weight (g/mol): 158.2
Molecular Formula: C8H14O3
InChi Key: LSACYLWPPQLVSM-UHFFFAOYSA-N
SMILES: CC(C)C(=O)OC(=O)C(C)C
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products