CAS RN 1076198-97-4
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
7-Benzyloxy-N-des[[2-(2-hydroxy)ethoxy]ethyl] Quetiapine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
7-Benzyloxy-N-des[[2-(2-hydroxy)ethoxy]ethyl] Quetiapine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
7-Benzyloxy-N-des[[2-(2-hydroxy)ethoxy]ethyl] Quetiapine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.