CAS RN 111248-01-2
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
(8S-cis)-8-Acetyl-10-[[(1,1-dimethylethyl)dimethylsilyl]oxy]-7,8,9,10-tetrahydro-6,8,11-trihydroxy-1, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(8S-cis)-8-Acetyl-10-[[(1,1-dimethylethyl)dimethylsilyl]oxy]-7,8,9,10-tetrahydro-6,8,11-trihydroxy-1-methoxy-5,12-naphthacenedione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.