CAS RN 115097-56-8
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
3,20-Disemicarbazone-17,21-dihydroxy-pregna-1,4-diene-3,11,20-trione 21-Acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3,20-Disemicarbazone-17,21-dihydroxy-pregna-1,4-diene-3,11,20-trione 21-Acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3,20-Disemicarbazone-17,21-dihydroxy-pregna-1,4-diene-3,11,20-trione 21-Acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.