CAS RN 1215006-08-8
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
4
av
4
Resultat
1
N-t-Butyl N-Deactyl Linezolid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-t-Butyl N-Deactyl Linezolid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-t-Butyl N-Deactyl Linezolid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
(5S)-5-[[(1,1-Dimethylethyl)amino]methyl]-3-[3-fluoro-4-(4-morpholinyl)phenyl]-2-oxazolidinone, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.