CAS RN 1216447-65-2
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
4-[4-[4-(Diphenylmethoxy-d5)-1-piperidinyl]-1-butyne]-Alpha,Alpha-dimethyl-benzeneacetic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-[4-[4-(Diphenylmethoxy-d5)-1-piperidinyl]-1-butyne]-Alpha,Alpha-dimethyl-benzeneacetic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.