CAS RN 1216892-47-5
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
5H,6H,7H,8H-Imidazo[1,2-a]pyridine-3-sulfonyl Chloride (~85%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5H,6H,7H,8H-Imidazo[1,2-a]pyridine-3-sulfonyl Chloride (~85%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5H,6H,7H,8H-Imidazo[1,2-a]pyridine-3-sulfonyl Chloride (~85%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.