CAS RN 1345697-50-8
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
2,6-Dideoxy-3,4-bis-O-[(1,1-dimethylethyl)diphenylsilyl]-D-ribo-hexose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,6-Dideoxy-3,4-bis-O-[(1,1-dimethylethyl)diphenylsilyl]-D-ribo-hexose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,6-Dideoxy-3,4-bis-O-[(1,1-dimethylethyl)diphenylsilyl]-D-ribo-hexose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.