CAS RN 15981-63-2
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
2’,3’,5’-Tri-O-acetylnebularine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2’,3’,5’-Tri-O-acetylnebularine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2’,3’,5’-Tri-O-acetylnebularine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.