CAS RN 1616761-00-2
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
N-Methyl-N-(3R,4R)-1-cyanoacetyl-4-methylpiperidin-3-yl-2-chloro-7-deazapurine-6-amine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Methyl-N-(3R,4R)-1-cyanoacetyl-4-methylpiperidin-3-yl-2-chloro-7-deazapurine-6-amine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.