CAS RN 1691234-08-8
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
(3Beta,5Alpha)-3-[[(1,1-Dimethylethyl)diphenylsilyl]oxy]-cholest-7-en-24-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(3b,5a)-3-[[(1,1-Dimethylethyl)diphenylsilyl]oxy]-cholest-7-en-24-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(3Beta,5Alpha)-3-[[(1,1-Dimethylethyl)diphenylsilyl]oxy]-cholest-7-en-24-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.