CAS RN 169528-22-7
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
N-tert-Butyloxycarbonyl-D-erythro-dihydro-D-sphingosine-1-phosphate Dicyanoethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-tert-Butyloxycarbonyl-D-erythro-dihydro-D-sphingosine-1-phosphate Dicyanoethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.