CAS RN 169738-14-1
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
(3R-cis)-Dihydro-3-hydroxy-4-(phenylmethoxy)-2(3H)-furanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(3R-cis)-Dihydro-3-hydroxy-4-(phenylmethoxy)-2(3H)-furanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(3R-cis)-Dihydro-3-hydroxy-4-(phenylmethoxy)-2(3H)-furanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.