CAS RN 172377-52-5
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
1
av
1
Resultat
1
Estradiol 3-sulfamate, MedChemExpress
MedChemExpress Estradiol 3-sulfamate (BLE 00084; E2MATE; ES-J 995) is a potent, long-acting, and orally active steroid sulfatase inhibitor; inhibits estrone sulfatase with an IC50 of 251 nM and a Ki of 133 nM.