CAS RN 1797124-71-0
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
7-Hydroxy-4,5,6,7-tetrahydro-imidazo[4,5,1-jk][1]benzazepin-2(1H)-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
7-Hydroxy-4,5,6,7-tetrahydro-imidazo[4,5,1-jk][1]benzazepin-2(1H)-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.