CAS RN 18514-46-0
CAS RN 18514-46-0
IUPAC Namn:
ethyl 2-[(triphenylmethyl)amino]acetate
Synonymer:
ethyl 2-[(triphenylmethyl)amino]acetate
Molekylvikt (g/mol):
345.44
Molekylär sammansättning:
C23H23NO2
InChi Key:
CAFYNMFIEFNYRA-UHFFFAOYSA-N
SMILES:
CCOC(=O)CNC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
av
1
Resultat
1
N-Tritylglycine ethyl ester, 98%, Thermo Scientific™
CAS: 18514-46-0 Molekylformel: C23H23NO2 Molekylvikt (g/mol): 345.442 MDL-nummer: MFCD00026893 InChI-nyckel: CAFYNMFIEFNYRA-UHFFFAOYSA-N Synonym: n-triphenylmethyl glycine ethyl ester,trt-gly-oet,ethyl n-tritylglycinate,n-tritylglycine ethyl ester,ethyl 2-tritylamino acetate,ethyl 2-triphenylmethyl amino acetate,acmc-20ak9z,n-trityl-glycine ethylester,ethyl 2-triphenylmethyl amino ethanoate,glycine, n-triphenylmethyl-, ethyl ester PubChem CID: 262774 IUPAC-namn: ethyl 2-(tritylamino)acetate LEDER: CCOC(=O)CNC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3