CAS RN 18856-86-5
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
4
av
4
Resultat
1
Trimethylamine-d9 Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Trimethyl-d9-amine HCl, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
3
Trimethylamine-d9 Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Trimethylamine-d9 Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.