CAS RN 190006-01-0
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
(3S,3aS,5aS,6R,9aS,9bS)-3-[[(1,1-Dimethylethyl)amino]carbonyl]dodecahydro-3a,6-dimethyl-7-oxo-1H-benz[e]indene-6-propanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-(1,1-Dimethylethyl)-17beta-carbamoyl-5-oxo-A-nor-3,5-secoandrostane-3-oic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.