CAS RN 197587-31-8
IUPAC Namn:
heptasodium {36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecahydroxy-10,15,20,25,30,35-hexakis[(sulfonatooxy)methyl]-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontan-5-yl}methyl sulfate
Synonymer:
heptasodium {36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecahydroxy-10,15,20,25,30,35-hexakis[(sulfonatooxy)methyl]-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontan-5-yl}methyl sulfate
Molekylvikt (g/mol):
1849.26
Molekylär sammansättning:
C42H63Na7O56S7
InChi Key:
TZBDYIAUDQVKGF-UHFFFAOYNA-G
SMILES:
[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].OC1C(O)C2OC3OC(COS([O-])(=O)=O)C(OC4OC(COS([O-])(=O)=O)C(OC5OC(COS([O-])(=O)=O)C(OC6OC(COS([O-])(=O)=O)C(OC7OC(COS([O-])(=O)=O)C(OC8OC(COS([O-])(=O)=O)C(OC1OC2COS([O-])(=O)=O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
1
–
3
av
3
Resultat
1
Heptakis(6-O-sulfo)-Beta-cyclodextrin Heptasodium Salt (>90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Heptakis(6-O-sulfo)-Beta-cyclodextrin Heptasodium Salt (>90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Heptakis(6-O-sulfo)-Beta-cyclodextrin Heptasodium Salt (>90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.