CAS RN 198637-59-1
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
7-[(5R,6S)-6-Hydroxy-2,2-dimethyl-1,3-dioxepan-5-yl]-1,4,7,10-tetraazacyclododecane-1-carboxaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
7-[(5R,6S)-6-Hydroxy-2,2-dimethyl-1,3-dioxepan-5-yl]-1,4,7,10-tetraazacyclododecane-1-carboxaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
7-[(5R,6S)-6-Hydroxy-2,2-dimethyl-1,3-dioxepan-5-yl]-1,4,7,10-tetraazacyclododecane-1-carboxaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.