CAS RN 2114-39-8
IUPAC Namn:
(2-bromopropyl)benzene
Synonymer:
(2-bromopropyl)benzene
Molekylvikt (g/mol):
199.09
Molekylär sammansättning:
C9H11Br
InChi Key:
NVYOCAOZCSNIHR-UHFFFAOYNA-N
SMILES:
CC(Br)CC1=CC=CC=C1
1
–
3
av
3
Resultat
1
2-Bromo-1-phenylpropane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Bromo-1-phenylpropane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Bromo-1-phenylpropane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.