CAS RN 2407569-85-9
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
(4Z,7Z,10Z,13Z,16Z,19Z)-N-[6-Hydroxy-5-(hydroxymethyl)hexyl]-4,7,10,13,16,19-docosahexaenamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(4Z,7Z,10Z,13Z,16Z,19Z)-N-[6-Hydroxy-5-(hydroxymethyl)hexyl]-4,7,10,13,16,19-docosahexaenamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(4Z,7Z,10Z,13Z,16Z,19Z)-N-[6-Hydroxy-5-(hydroxymethyl)hexyl]-4,7,10,13,16,19-docosahexaenamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.