CAS RN 244056-98-2
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
(3aS*,7aS*)-Benzyl 2-(bromomethyl)hexahydrofuro[3,2-b]pyridine-4(2H)-carboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(3aS*,7aS*)-Benzyl 2-(bromomethyl)hexahydrofuro[3,2-b]pyridine-4(2H)-carboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(3aS*,7aS*)-Benzyl 2-(bromomethyl)hexahydrofuro[3,2-b]pyridine-4(2H)-carboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.