CAS RN 290295-98-6
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
1
av
1
Resultat
1
(2-Butyl-3-benzofuranyl)(4-Ethoxy-3,5-diiodophenyl)methanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.