CAS RN 3282-99-3
IUPAC Namn:
4-[4-(4-aminophenyl)cyclohexyl]aniline
Synonymer:
4-[4-(4-aminophenyl)cyclohexyl]aniline
Molekylvikt (g/mol):
266.39
Molekylär sammansättning:
C18H22N2
InChi Key:
DJCVCYUEICJZBD-UHFFFAOYSA-N
SMILES:
NC1=CC=C(C=C1)C1CCC(CC1)C1=CC=C(N)C=C1
1
–
3
av
3
Resultat
1
1,1-Bis(4-aminophenyl)cyclohexane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,1-Bis(4-aminophenyl)cyclohexane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1,1-Bis(4-aminophenyl)cyclohexane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.