CAS RN 3330-14-1
IUPAC Namn:
1,1,1,2,2,3,3-heptafluoro-3-{[1,1,1,2,3,3-hexafluoro-3-(1,2,2,2-tetrafluoroethoxy)propan-2-yl]oxy}propane
Synonymer:
1,1,1,2,2,3,3-heptafluoro-3-{[1,1,1,2,3,3-hexafluoro-3-(1,2,2,2-tetrafluoroethoxy)propan-2-yl]oxy}propane
Molekylvikt (g/mol):
452.07
Molekylär sammansättning:
C8HF17O2
InChi Key:
PYSYKOPZHYNYSZ-UHFFFAOYNA-N
SMILES:
FC(OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F)C(F)(F)F