CAS RN 3979-51-9
IUPAC Namn:
propan-2-(²H)ol
Synonymer:
propan-2-(²H)ol
Molekylvikt (g/mol):
61.1
Molekylär sammansättning:
C3H8O
InChi Key:
KFZMGEQAYNKOFK-QYKNYGDISA-N
SMILES:
[2H]OC(C)C
1
–
1
av
1
Resultat
1
iso-Propyl Alcohol-OD, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.