CAS RN 41263-74-5
IUPAC Namn:
4-(methylamino)-3-nitrobenzoate
Synonymer:
4-(methylamino)-3-nitrobenzoate
Molekylvikt (g/mol):
195.16
Molekylär sammansättning:
C8H7N2O4
InChi Key:
KSMLIIWEQBYUKA-UHFFFAOYSA-M
SMILES:
CNC1=CC=C(C=C1[N+]([O-])=O)C([O-])=O
1
–
3
av
3
Resultat
1
4-(Methylamino)-3-nitrobenzoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-(Methylamino)-3-nitrobenzoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-(Methylamino)-3-nitrobenzoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.