CAS RN 443103-97-7
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
1
av
1
Resultat
1
A2A receptor antagonist 1, MedChemExpress
MedChemExpress A2A receptor antagonist 1 (CPI-444 analog) is an antagonist of both adenosine A2A receptor and A1 receptor with Ki values of 4 and 264 nM, respectively.